Assay Details
Target Gene Details
Entrez Gene ID: | 68778 |
Gene Name: | guanylyl cyclase domain containing 1 |
Gene Aliases: |
1110038D17Rik |
Location: |
Chr.10:75506814-75517967 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucd1 | NM_175133.1 | NP_780342.1 | ||
| AK088820.1 | ||||
| AK157404.1 | BAE34080.1 | |||
| AK170872.1 | ||||
| BC024851.1 | ||||
| BC043463.1 | ||||
| BC118613.1 | ||||
| BC119795.1 |
More Information
Set Membership: |
|

Genomic Map