Assay Details
Target Gene Details
Entrez Gene ID: | 11776 |
Gene Name: | adaptor-related protein complex 3, delta 1 subunit |
Gene Aliases: |
AA407035, Ap3d, Bolvr, mBLVR1, mh, mocha |
Location: |
Chr.10:80706956-80742282 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap3d1 | NM_007460.1 | 14 | 1811 | NP_031486.1 |
| AB004305.1 | 14 | 1811 | BAA24578.1 | |
| AK030627.1 | 1 | 1166 | ||
| AK132564.1 | 1 | 213 | ||
| AK141789.1 | 14 | 1659 | BAE24834.1 | |
| AK155518.1 | 14 | 1861 | BAE33305.1 | |
| AK199246.1 | ||||
| AK204130.1 | ||||
| BC022171.1 | 14 | 1816 | ||
| BC037477.1 | 14 | 1812 | AAH37477.1 | |
| BC048786.1 | 14 | 1801 | AAH48786.1 | |
| BC053066.1 | 14 | 1827 | AAH53066.1 |
More Information
Set Membership: |
|

Genomic Map