Assay Details
Target Gene Details
Entrez Gene ID: | 70574 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
1110060I01Rik, 5730456K23Rik, AA589379, E030045M14Rik |
Location: |
Chr.10:117629500-117687352 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpm | NM_027468.1 | 5 | 556 | NP_081744.1 |
| AK087332.1 | 1 | 917 | ||
| BC047389.1 | 4 | 542 | AAH47389.1 | |
| BC100404.1 | 5 | 550 | AAI00405.1 |
More Information
Set Membership: |
|

Genomic Map