Assay Details
Target Gene Details
Entrez Gene ID: | 12905 |
Gene Name: | CASP2 and RIPK1 domain containing adaptor with death domain |
Gene Aliases: |
RAIDD |
Location: |
Chr.10:95174739-95324345 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cradd | NM_009950.2 | NP_034080.1 | ||
AJ224738.1 | CAA12107.1 | |||
AK144648.1 | BAE25988.1 | |||
AK163112.1 | BAE37198.1 |
