Assay Details
Target Gene Details
Entrez Gene ID: | 67239 |
Gene Name: | ribosome production factor 2 homolog (S. cerevisiae) |
Gene Aliases: |
2810470K21Rik, AU040229, AU043242, Bxdc1 |
Location: |
Chr.10:40223246-40247039 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpf2 | NM_001042556.1 | NP_001036021.1 | ||
NM_023323.3 | NP_075812.3 | |||
AB041810.1 | BAA95117.1 | |||
AK010776.1 | BAB27175.1 | |||
AK011644.1 | ||||
AK012188.1 | BAB28087.1 | |||
AK012641.1 | BAB28375.1 | |||
AK013395.1 | BAB28829.1 | |||
AK017678.1 | BAB30868.1 | |||
AK020030.1 | BAB31973.1 | |||
AK051135.1 |
