Assay Details
Target Gene Details
Entrez Gene ID: | 78334 |
Gene Name: | cyclin-dependent kinase 19 |
Gene Aliases: |
2700084L06Rik, AI845279, AW228747, CDK11, Cdc2l6, mKIAA1028 |
Location: |
Chr.10:40349308-40483818 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk19 | NM_001168304.1 | NP_001161776.1 | ||
| NM_001291816.1 | NP_001278745.1 | |||
| NM_001291817.1 | NP_001278746.1 | |||
| NM_198164.3 | NP_937807.2 | |||
| AK052818.1 | BAC35159.1 | |||
| AK053651.1 | BAC35463.1 | |||
| AK122421.1 | BAC65703.1 | |||
| AK147340.1 | ||||
| BC030312.1 | AAH30312.1 | |||
| BC137614.1 | ||||
| BC144782.1 |
More Information
Set Membership: |
|

Genomic Map