Assay Details
Target Gene Details
Entrez Gene ID: | 56530 |
Gene Name: | canopy 2 homolog (zebrafish) |
Gene Aliases: |
5330432A10Rik, AW229003, D10Bwg1546e, Tmem4, Zsig9 |
Location: |
Chr.10:128322431-128327187 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnpy2 | NM_019953.1 | NP_064337.1 | ||
AF186115.1 | AAF01433.1 | |||
AK007914.1 | BAB25346.1 | |||
AK013014.1 | BAB28597.1 | |||
AK013568.1 | BAB28909.1 | |||
AK019927.1 | BAB31921.1 | |||
AK150953.1 | BAE29986.1 | |||
BC008261.1 | AAH08261.1 |
