Assay Details
Target Gene Details
Entrez Gene ID: | 11702 |
Gene Name: | S-adenosylmethionine decarboxylase 1 |
Gene Aliases: |
1, AdoMetDC, Amd-1, SAMDC, SAMDC 1, adoMetDC1 |
Location: |
Chr.10:40287458-40302188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amd1 | NM_009665.5 | NP_033795.1 | ||
| AK146840.1 | BAE27473.1 | |||
| AK166187.1 | BAE38617.1 | |||
| AK168165.1 | BAE40126.1 | |||
| AK171238.1 | ||||
| BC011110.1 | AAH11110.1 | |||
| BC071220.1 | AAH71220.1 | |||
| BC080791.1 | AAH80791.1 | |||
| BC092072.1 | AAH92072.1 | |||
| BC138696.1 | ||||
| BC138697.1 | ||||
| CT010192.1 | CAJ18400.1 | |||
| D12780.1 | BAA02243.1 | |||
| Z14986.1 | CAA78710.1 |
More Information
Set Membership: |
|

Genomic Map