Assay Details
Target Gene Details
Entrez Gene ID: | 103236 |
Gene Name: | casein kinase 1, gamma 2 |
Gene Aliases: |
2810429I12Rik, AI463719 |
Location: |
Chr.10:80622705-80640771 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1g2 | NM_001159591.1 | 6 | 869 | NP_001153063.1 |
| NM_134002.2 | 5 | 1046 | NP_598763.1 | |
| AK077076.1 | 6 | 849 | BAC36596.1 | |
| AK132871.1 | 6 | 870 | BAE21399.1 | |
| AK152413.1 | 5 | 1045 | ||
| BC004839.1 | 5 | 988 | AAH04839.1 | |
| BC005750.1 | AAH05750.1 |
More Information
Set Membership: |
|

Genomic Map