Assay Details
Target Gene Details
Entrez Gene ID: | 75317 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
4930547N16Rik, AI503644, PARI |
Location: |
Chr.10:88091398-88146945 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Parpbp | NM_029249.2 | 10 | 1778 | NP_083525.1 |
AK030970.1 | 5 | 607 | BAC27197.1 | |
AK131981.1 | 10 | 1764 | BAE20920.1 | |
AK137160.1 | 10 | 1727 | ||
AK150571.1 | 10 | 1742 | BAE29666.1 | |
AK151268.1 | 10 | 1742 | BAE30255.1 | |
AK152208.1 | 10 | 1742 | BAE31036.1 | |
BC070476.1 | 10 | 1778 |
