Assay Details
Target Gene Details
Entrez Gene ID: | 11783 |
Gene Name: | apoptotic peptidase activating factor 1 |
Gene Aliases: |
6230400I06Rik, Apaf-1, Apaf1l, fog, mKIAA0413 |
Location: |
Chr.10:90989311-91082743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apaf1 | NM_001042558.1 | NP_001036023.1 | ||
| NM_001282947.1 | NP_001269876.1 | |||
| NM_009684.2 | NP_033814.2 | |||
| AF064071.1 | AAC62458.1 | |||
| AK220340.1 | BAD90406.1 | |||
| BC131683.1 | ||||
| BC131684.1 |

Genomic Map