Assay Details
Target Gene Details
Entrez Gene ID: | 110532 |
Gene Name: | adenosine deaminase, RNA-specific, B1 |
Gene Aliases: |
1700057H01Rik, AW124433, AW558573, Adar2, BB220382, D10Bwg0447e, Red1 |
Location: |
Chr.10:77290727-77418273 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adarb1 | NM_001024837.2 | 10 | 2303 | NP_001020008.1 |
| NM_130895.3 | 10 | 2273 | NP_570965.2 | |
| NR_004429.1 | 10 | 2350 | ||
| NR_021486.1 | 10 | 2320 | ||
| AF291049.1 | 9 | 2108 | AAK17102.1 | |
| AF403106.2 | 10 | 2225 | AAL01301.1 | |
| AF403107.2 | 10 | 2255 | AAL01302.1 | |
| AF403108.2 | 10 | 2272 | AAL01303.1 | |
| AF403109.2 | 10 | 2302 | AAL01304.1 | |
| AF525421.1 | 11 | 2298 | AAM83099.1 | |
| AK139004.1 | 1 | 105 | BAE23858.1 | |
| AK141777.1 | 10 | 2275 | BAE24831.1 | |
| AK147298.1 | 11 | 2382 | BAE27830.1 | |
| AY162454.1 | 11 | 2298 | AAN86546.1 |
More Information
Set Membership: |
|

Genomic Map