Assay Details
Target Gene Details
Entrez Gene ID: | 57267 |
Gene Name: | amyloid beta (A4) precursor protein-binding, family A, member 3 |
Gene Aliases: |
AW208791, Mint-3, X11gamma, lin-10, mint3 |
Location: |
Chr.10:81266965-81273247 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apba3 | NM_018758.2 | NP_061228.1 | ||
| AF070975.1 | AAC78837.1 | |||
| AK045965.1 | BAC32549.1 | |||
| AK169462.1 | BAE41187.1 | |||
| AK170182.1 | BAE41620.1 | |||
| BC005423.1 | AAH05423.1 | |||
| BC009666.1 | AAH09666.1 |
Target Gene Details
Entrez Gene ID: | 100526465 |
Gene Name: | microRNA 3057 |
Gene Aliases: |
mir-3057, mmu-mir-3057 |
Location: |
Chr.10:81271597-81271687 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir3057 | NR_037218.1 |
More Information
Set Membership: |
|

Genomic Map