Assay Details
Target Gene Details
Entrez Gene ID: | 16467 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human) |
Gene Aliases: |
3322401A10Rik, BB077577, BNIP-H, CLAC, hes, ji |
Location: |
Chr.10:81204513-81230779 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atcay | NM_178662.3 | NP_848777.1 | ||
| AK028298.1 | BAC25867.1 | |||
| AK038483.1 | BAC30014.1 | |||
| AK038990.1 | BAC30195.1 | |||
| AK039411.1 | BAC30342.1 | |||
| AK086723.1 | ||||
| AK162959.1 | BAE37136.1 | |||
| AY349150.1 | AAQ90064.1 | |||
| BC048903.1 | AAH48903.1 |
Target Gene Details
Entrez Gene ID: | 69678 |
Gene Name: | ataxia, cerebellar, Cayman type homolog (human), opposite strand |
Gene Aliases: |
2310050B05Rik |
Location: |
Chr.10:81194691-81208370 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atcayos | NR_015477.1 | |||
| AK084504.1 |
More Information
Set Membership: |
|

Genomic Map