Assay Details
Target Gene Details
Entrez Gene ID: | 110279 |
Gene Name: | breakpoint cluster region |
Gene Aliases: |
5133400C09Rik, AI561783, AI853148, mKIAA3017 |
Location: |
Chr.10:75060557-75184923 on Build GRCm38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcr | NM_001081412.2 | NP_001074881.1 | ||
| AK017213.1 | ||||
| AK129482.1 | BAC98292.1 | |||
| BC048842.1 | AAH48842.1 | |||
| BC060270.1 | AAH60270.1 | |||
| BC131682.1 |
More Information
Set Membership: |
|

Genomic Map