Assay Details
Target Gene Details
Entrez Gene ID: | 216439 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
AGAP-2, Centg1, PIKE, cnt-g1, mKIAA0167 |
Location: |
Chr.10:127075258-127093173 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agap2 | NM_001033263.5 | NP_001028435.1 | ||
| NM_001301014.1 | NP_001287943.1 | |||
| AK147446.1 | BAE27918.1 | |||
| AK220325.1 | BAD90236.1 |

Genomic Map