Assay Details
Target Gene Details
Entrez Gene ID: | 216190 |
Gene Name: | adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2 |
Gene Aliases: |
Dip3b |
Location: |
Chr.10:83600033-83648854 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Appl2 | NM_145220.2 | NP_660255.1 | ||
| AK160075.1 | BAE35609.1 | |||
| AK160105.1 | BAE35632.1 | |||
| AK168403.1 | ||||
| AK168748.1 | BAE40588.1 | |||
| AK171512.1 | BAE42499.1 | |||
| AY113706.1 | AAM55532.1 | |||
| BC002232.1 | AAH02232.1 | |||
| BC048906.1 | AAH48906.1 |
More Information
Set Membership: |
|

Genomic Map