Assay Details
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Exon 51 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh23 | NM_001252635.1 | 50 | 7310 | NP_001239564.1 |
| NM_023370.3 | 51 | 7316 | NP_075859.2 | |
| NR_045556.1 | 50 | 7310 | ||
| AF308939.1 | 49 | 6909 | AAG52817.1 | |
| AJ888000.1 | 48 | 6903 | CAI59609.1 | |
| AJ888001.1 | 48 | 6903 | CAI59610.1 | |
| AJ888002.1 | 48 | 6903 | CAI59611.1 | |
| AK039126.1 | 6 | 651 | ||
| AY026062.1 | 49 | 6918 | AAK07670.1 | |
| AY563163.1 | 3 | 531 | AAT72163.1 | |
| AY563164.1 | 3 | 531 | AAT72164.1 |
More Information
Set Membership: |
|

Genomic Map