Assay Details
Target Gene Details
Entrez Gene ID: | 11947 |
Gene Name: | ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit |
Gene Aliases: |
- |
Location: |
Chr.10:128083307-128090388 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
More Information
Set Membership: |
|

Genomic Map