Assay Details
Target Gene Details
Entrez Gene ID: | 12822 |
Gene Name: | collagen, type XVIII, alpha 1 |
Gene Aliases: |
- |
Location: |
Chr.10:77052178-77166530 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col18a1 | NM_001109991.1 | NP_001103461.1 | ||
| NM_009929.3 | NP_034059.2 | |||
| AK031798.1 | BAC27554.1 | |||
| BC064817.1 | AAH64817.1 | |||
| BC066080.1 | AAH66080.1 | |||
| D17546.1 | BAA04483.1 | |||
| L16898.1 | AAA37434.1 | |||
| L22545.1 | AAA19787.1 |
More Information
Set Membership: |
|

Genomic Map