Assay Details
Target Gene Details
Entrez Gene ID: | 73910 |
Gene Name: | Rho GTPase activating protein 18 |
Gene Aliases: |
4833419J07Rik, AI314906, AI843501 |
Location: |
Chr.10:26695312-26918648 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap18 | NM_176837.2 | NP_789807.1 | ||
| AK014728.1 | ||||
| AK078522.1 | BAC37322.1 | |||
| AK087800.1 | ||||
| AK162202.1 | ||||
| BC030858.1 | AAH30858.1 |
More Information
Set Membership: |
|

Genomic Map