Assay Details
Target Gene Details
Entrez Gene ID: | 13202 |
Gene Name: | D-dopachrome tautomerase |
Gene Aliases: |
C78655 |
Location: |
Chr.10:75771233-75773374 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddt | NM_010027.1 | NP_034157.1 | ||
| AK144193.1 | BAE25759.1 | |||
| BC010753.1 | AAH10753.1 |
More Information
Set Membership: |
|

Genomic Map