Assay Details
Target Gene Details
Entrez Gene ID: | 56200 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 21 |
Gene Aliases: |
AI255159, AL022742, D10Ertd645e, D10Wsu42e |
Location: |
Chr.10:62580247-62602298 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx21 | NM_019553.2 | 1 | 128 | NP_062426.2 |
AF159131.3 | 1 | 89 | AAD43959.3 | |
AK010281.1 | BAB26817.2 | |||
AK012125.1 | 1 | 85 | BAB28047.2 | |
AK145578.1 | 1 | 112 | BAE26522.1 | |
AK160095.1 | 1 | 102 | BAE35625.1 | |
BC043655.1 | 1 | 100 | ||
BC059237.1 | 1 | 88 | AAH59237.1 | |
CF182162.1 | 1 | 145 |
