Assay Details
Target Gene Details
Entrez Gene ID: | 216445 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
AU043488 |
Location: |
Chr.10:127321959-127329946 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap9 | NM_001285785.1 | NP_001272714.1 | ||
| NM_146011.2 | NP_666123.2 | |||
| AK047280.1 | BAC33014.1 | |||
| AK049245.1 | 1 | 101 | BAC33633.1 | |
| AK080955.1 | BAC38093.1 | |||
| AK083474.1 | 1 | 230 | ||
| AK089051.1 | BAC40720.1 | |||
| AK167738.1 | BAE39777.1 | |||
| BC024535.1 | ||||
| BC027374.1 | ||||
| BY184139.1 | ||||
| DQ498128.1 |
More Information
Set Membership: |
|

Genomic Map