Assay Details
Target Gene Details
Entrez Gene ID: | 76138 |
Gene Name: | coiled-coil domain containing 138 |
Gene Aliases: |
6230424H07Rik, BC042726 |
Location: |
Chr.10:58497701-58576244 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc138 | NM_001162956.1 | 4 | 286 | NP_001156428.1 |
BC119028.1 | 4 | 275 | ||
BC119030.1 | 4 | 275 |
