Assay Details
Target Gene Details
Entrez Gene ID: | 216443 |
Gene Name: | methionine-tRNA synthetase |
Gene Aliases: |
Metrs, Mtrns |
Location: |
Chr.10:127296221-127311786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mars | NM_001003913.2 | NP_001003913.1 | ||
| NM_001171582.1 | NP_001165053.1 | |||
| AK190213.1 | ||||
| AK191034.1 | ||||
| AK198349.1 | ||||
| AK201611.1 | ||||
| BC079643.1 | AAH79643.1 |
More Information
Set Membership: |
|

Genomic Map