Assay Details
Target Gene Details
Entrez Gene ID: | 171580 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 1 |
Gene Aliases: |
MICAL, MICAL-1, Nical |
Location: |
Chr.10:41475860-41487030 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mical1 | NM_001164433.1 | 16 | 2278 | NP_001157905.1 |
NM_138315.2 | 17 | 2497 | NP_612188.1 | |
AK154665.1 | 17 | 2437 | ||
AK154930.1 | 17 | 2477 | ||
AK157196.1 | 17 | 2434 | ||
AK159369.1 | 16 | 2225 | BAE35027.1 | |
AK170382.1 | 17 | 2437 | ||
AK170827.1 | 17 | 2469 | ||
AK170874.1 | 17 | 2479 | ||
AK170881.1 | 17 | 2463 | BAE42090.1 | |
AK171234.1 | 17 | 2464 | BAE42330.1 | |
AK171429.1 | 17 | 2476 | BAE42447.1 | |
AK171479.1 | 17 | 2470 | BAE42481.1 | |
AK171779.1 | 14 | 3429 | ||
AK172562.1 | 17 | 2471 | ||
AK172716.1 | 17 | 2423 | ||
AK185820.1 | 3 | 275 | ||
AK206362.1 | 3 | 254 | ||
BC021477.1 | 17 | 2445 | AAH21477.1 | |
BC034682.1 | 17 | 2389 | AAH34682.1 |
