Assay Details
Target Gene Details
Entrez Gene ID: | 69912 |
Gene Name: | nucleoporin 43 |
Gene Aliases: |
2610016K01Rik, 2610529I12Rik, AA409950, p42 |
Location: |
Chr.10:7667504-7678886 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nup43 | NM_145706.2 | 5 | 622 | NP_663752.2 |
AK011422.1 | 5 | 587 | ||
AK012191.1 | 5 | 615 | BAC25359.1 | |
AK090019.1 | 5 | 585 | BAC41049.1 | |
AK141339.1 | 5 | 587 | BAE24655.1 | |
AK146262.1 | 5 | 624 | BAE27021.1 | |
BC011060.1 | 5 | 610 | AAH11060.1 | |
BC052530.1 | 5 | 550 | AAH52530.2 | |
BF607996.1 | 4 | 390 |
Target Gene Details
Entrez Gene ID: | 18537 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase 1 |
Gene Aliases: |
C79501, PIMT |
Location: |
Chr.10:7629380-7680973 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcmt1 |
