Assay Details
Target Gene Details
Entrez Gene ID: | 69912 |
Gene Name: | nucleoporin 43 |
Gene Aliases: |
2610016K01Rik, 2610529I12Rik, AA409950, p42 |
Location: |
Chr.10:7667504-7678886 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nup43 | NM_145706.2 | NP_663752.2 | ||
AK011422.1 | ||||
AK012191.1 | BAC25359.1 | |||
AK090019.1 | BAC41049.1 | |||
AK141339.1 | 6 | 785 | BAE24655.1 | |
AK146262.1 | BAE27021.1 | |||
BC011060.1 | AAH11060.1 | |||
BC052530.1 | AAH52530.2 | |||
BF607996.1 |
Target Gene Details
Entrez Gene ID: | 18537 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase 1 |
Gene Aliases: |
C79501, PIMT |
Location: |
Chr.10:7629380-7680973 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcmt1 |
