Assay Details
Target Gene Details
Entrez Gene ID: | 100316778 |
Gene Name: | microRNA 1982 |
Gene Aliases: |
mir-1982, mmu-mir-1982 |
Location: |
Chr.10:80828797-80828870 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir1982 | NR_035503.1 |
Target Gene Details
Entrez Gene ID: | 18245 |
Gene Name: | ornithine decarboxylase antizyme 1 |
Gene Aliases: |
AZ-1, AZ1, Antizyme, ODC-Az, Oaz |
Location: |
Chr.10:80826656-80829290 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oaz1 | NM_001301034.1 | NP_001287963.1 | ||
| NM_008753.4 | NP_032779.2 | |||
| AK003452.1 | 4 | 616 | ||
| AK010615.1 | ||||
| AK019155.1 | ||||
| AK087942.1 | 2 | 2029 | ||
| AK146571.1 | ||||
| AK147167.1 | ||||
| AK151635.1 | ||||
| AK159849.1 | ||||
| AK159874.1 | ||||
| AK160491.1 | ||||
| AK160598.1 | ||||
| AK169201.1 | ||||
| AK201392.1 | ||||
| BC094287.1 | AAH94287.1 | |||
| BG296334.1 | ||||
| U52822.1 | AAB96329.1 |
More Information
Set Membership: |
|

Genomic Map