Assay Details
Target Gene Details
Entrez Gene ID: | 432486 |
Gene Name: | N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits |
Gene Aliases: |
EG432486, mKIAA1208 |
Location: |
Chr.10:88379137-88447330 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnptab | NM_001004164.2 | 10 | 1196 | NP_001004164.2 |
AK140867.1 | 9 | 1300 | BAE24503.1 | |
AK154710.1 | 2 | 95 | BAE32779.1 | |
AK173132.1 | 10 | 1196 | BAD32410.1 | |
BC060638.1 | 11 | 1304 | AAH60638.1 |
