Assay Details
Target Gene Details
Entrez Gene ID: | 407790 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2 |
Gene Aliases: |
BC064011 |
Location: |
Chr.10:127509304-127517154 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa4l2 | NM_001098789.1 | NP_001092259.1 | ||
| AK131609.1 | BAE20718.1 | |||
| AK131662.1 | BAE20749.1 | |||
| BC099496.1 | AAH99496.1 | |||
| BY336656.1 |
More Information
Set Membership: |
|

Genomic Map