Assay Details
Target Gene Details
Entrez Gene ID: | 13629 |
Gene Name: | eukaryotic translation elongation factor 2 |
Gene Aliases: |
Ef-2 |
Location: |
Chr.10:81176631-81182509 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
More Information
Set Membership: |
|

Genomic Map