Assay Details
Target Gene Details
Entrez Gene ID: | 17319 |
Gene Name: | macrophage migration inhibitory factor |
Gene Aliases: |
GIF, Glif |
Location: |
Chr.10:75859353-75860250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mif | NM_010798.2 | NP_034928.1 | ||
| AK008939.1 | BAB25980.1 | |||
| AK010691.1 | BAB27123.1 | |||
| AK013328.1 | BAB28792.1 | |||
| AK152211.1 | BAE31039.1 | |||
| AK218046.1 | ||||
| BC024895.1 | AAH24895.1 | |||
| BC086928.1 | AAH86928.1 | |||
| BC100605.1 | AAI00606.1 | |||
| L02913.1 | ||||
| L07607.1 | AAA37111.1 | |||
| L10613.1 | AAA37693.1 | |||
| Z23048.1 | CAA80583.1 |
More Information
Set Membership: |
|

Genomic Map