Assay Details
Target Gene Details
Entrez Gene ID: | 66844 |
Gene Name: | ORM1-like 2 (S. cerevisiae) |
Gene Aliases: |
0610012C09Rik |
Location: |
Chr.10:128817457-128821632 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ormdl2 | NM_024180.5 | NP_077142.1 | ||
BC002146.1 | AAH02146.1 | |||
DV065903.1 |
