Assay Details
Target Gene Details
Entrez Gene ID: | 15353 |
Gene Name: | high mobility group 20B |
Gene Aliases: |
AW610687, BRAF35, Hmgx2, Hmgxb2, Smarce1r |
Location: |
Chr.10:81346046-81350511 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmg20b | NM_001163165.1 | NP_001156637.1 | ||
| NM_001163166.1 | NP_001156638.1 | |||
| NM_010440.3 | NP_034570.1 | |||
| AF146224.1 | AAF66708.1 | |||
| AK153816.1 | BAE32194.1 | |||
| AK155652.1 | BAE33368.1 | |||
| AK155890.1 | BAE33486.1 | |||
| AL355735.1 | CAB90814.1 | |||
| BC003924.1 | ||||
| BC011334.1 | AAH11334.1 |

Genomic Map