Assay Details
Target Gene Details
Entrez Gene ID: | 23943 |
Gene Name: | extended synaptotagmin-like protein 1 |
Gene Aliases: |
Fam62a, Mbc2, vp115 |
Location: |
Chr.10:128510248-128525859 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esyt1 | NM_011843.2 | NP_035973.1 | ||
AF098633.1 | AAD10189.1 | |||
AF098634.1 | AAD10190.1 | |||
AK008224.1 | BAB25542.1 | |||
AK054147.1 | ||||
AK088421.1 | ||||
AK181044.1 | ||||
BC011482.1 | AAH11482.1 |
