Assay Details
Target Gene Details
Entrez Gene ID: | 215798 |
Gene Name: | adhesion G protein-coupled receptor G6 |
Gene Aliases: |
1190004A11Rik, AI449247, AW045736, DREG, Gm222, Gpr126 |
Location: |
Chr.10:14402580-14545722 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgrg6 | NM_001002268.3 | NP_001002268.1 | ||
| AB183545.1 | BAD27570.1 | |||
| BC046534.1 | AAH46534.1 | |||
| BC080779.1 | AAH80779.1 |
More Information
Set Membership: |
|

Genomic Map