Assay Details
Target Gene Details
Entrez Gene ID: | 208228 |
Gene Name: | MOB kinase activator 3A |
Gene Aliases: |
5330417K06Rik, A630029F06, AV218468, Mobkl2a |
Location: |
Chr.10:80685253-80701896 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob3a | NM_172457.2 | NP_766045.2 | ||
| AK030481.1 | BAC26983.1 | |||
| AK080169.1 | ||||
| AK170394.1 | BAE41766.1 | |||
| BC058238.1 | AAH58238.1 |
More Information
Set Membership: |
|

Genomic Map