Assay Details
Target Gene Details
Entrez Gene ID: | 209558 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 3 |
Gene Aliases: |
AI876438, CD203c |
Location: |
Chr.10:24773814-24836195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp3 | NM_134005.2 | NP_598766.2 | ||
AK089553.1 | 2 | 761 | ||
AK132256.1 | ||||
AY630402.1 | AAT64421.1 | |||
BC099535.1 | AAH99535.1 |
