Assay Details
Target Gene Details
Entrez Gene ID: | 23887 |
Gene Name: | gamma-glutamyltransferase 5 |
Gene Aliases: |
AI551243, GGL, GGT-REL, Ggtla1 |
Location: |
Chr.10:75589327-75616968 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ggt5 | NM_011820.4 | 10 | 1725 | NP_035950.2 |
AF077765.1 | 10 | 1710 | AAC71001.1 | |
AK029138.1 | 3 | 261 | BAC26318.1 | |
AK049084.1 | 10 | 1729 | ||
AK052183.1 | 10 | 1726 | BAC34873.1 | |
AK140308.1 | 9 | 2246 | ||
AK170326.1 | 10 | 1589 | BAE41720.1 | |
BC051971.1 | 9 | 1420 | AAH51971.1 | |
BC113775.1 | 10 | 1449 |
