Assay Details
Target Gene Details
Entrez Gene ID: | 13998 |
Gene Name: | FYVE, RhoGEF and PH domain containing 6 |
Gene Aliases: |
AA123052, Etohd4, ZFYVE24 |
Location: |
Chr.10:94036001-94145339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgd6 | NM_053072.3 | NP_444302.4 | ||
| AK016940.1 | BAB30510.2 | |||
| AK044341.1 | BAC31876.1 | |||
| AK173157.1 | BAD32435.1 | |||
| BC026860.1 | ||||
| BC139019.1 | ||||
| BC139022.1 |
More Information
Set Membership: |
|

Genomic Map