Assay Details
Target Gene Details
Entrez Gene ID: | 14632 |
Gene Name: | GLI-Kruppel family member GLI1 |
Gene Aliases: |
AV235269, Zfp-5, Zfp5 |
Location: |
Chr.10:127329881-127341579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gli1 | NM_010296.2 | NP_034426.2 | ||
| AB025922.1 | BAA85004.1 | |||
| AB232675.1 | ||||
| AB234616.1 | ||||
| AB234617.1 | ||||
| AF026305.1 | AAC09169.1 | |||
| AK157048.1 | ||||
| BC131649.1 | ||||
| BC131650.1 | ||||
| CJ062804.1 |
More Information
Set Membership: |
|

Genomic Map