Assay Details
Target Gene Details
Entrez Gene ID: | 432450 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
6330571D19Rik, AW455467, Tcba1 |
Location: |
Chr.10:31689310-32893997 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nkain2 | NM_001013411.2 | NP_001013429.2 | ||
AY940478.1 | AAX33293.1 | |||
DQ065607.1 | AAY68416.1 | |||
EF058049.2 | ||||
EF058050.1 |
