Assay Details
Target Gene Details
Entrez Gene ID: | 432450 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
6330571D19Rik, AW455467, Tcba1 |
Location: |
Chr.10:31689310-32893997 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkain2 | NM_001013411.2 | NP_001013429.2 | ||
| NM_001025286.2 | NP_001020457.2 | |||
| AY940478.1 | AAX33293.1 | |||
| BC141063.1 | ||||
| DQ065606.1 | AAY68415.1 | |||
| DQ065607.1 | AAY68416.1 | |||
| EF058049.2 | ||||
| EF058050.1 | ||||
| EF058051.1 |

Genomic Map