Assay Details
Target Gene Details
Entrez Gene ID: | 237362 |
Gene Name: | neuropeptide FF receptor 1 |
Gene Aliases: |
Gm236, Gpr147, NPFF1, OT7T022 |
Location: |
Chr.10:61595484-61628762 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npffr1 |

Genomic Map