Assay Details
Target Gene Details
Entrez Gene ID: | 14806 |
Gene Name: | glutamate receptor, ionotropic, kainate 2 (beta 2) |
Gene Aliases: |
AW124492, GluK2, Glur-6, Glur6, Glurbeta2 |
Location: |
Chr.10:49094842-49789250 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grik2 | NM_001111268.1 | 12 | 2010 | NP_001104738.1 |
| NM_010349.2 | 12 | 2010 | NP_034479.2 | |
| BC131639.1 | 11 | 1691 | ||
| BC131640.1 | 11 | 1691 | ||
| D10054.1 | 12 | 1923 | BAA00943.1 | |
| X66117.1 | 11 | 1747 | CAA46907.1 |
More Information
Set Membership: |
|

Genomic Map