Assay Details
Target Gene Details
Entrez Gene ID: | 17245 |
Gene Name: | transformed mouse 3T3 cell double minute 1 |
Gene Aliases: |
Arrd2, Mdm-1 |
Location: |
Chr.10:118141537-118168999 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdm1 | NM_001162904.1 | NP_001156376.1 | ||
| NM_001162905.1 | NP_001156377.1 | |||
| NM_010785.2 | NP_034915.2 | |||
| NM_148922.3 | NP_683724.2 | |||
| AK004789.2 | BAB23566.1 | |||
| AK011769.1 | BAB27830.1 | |||
| BC137608.1 | ||||
| BC144777.1 |

Genomic Map