Assay Details
Target Gene Details
Entrez Gene ID: | 74330 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 14 |
Gene Aliases: |
5730551F12Rik, DNAJ, DRIP78, HDJ3, LIP6 |
Location: |
Chr.10:128805468-128819446 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc14 | NM_028873.4 | NP_083149.3 | ||
| AK017830.1 | BAB30962.1 | |||
| AK032681.1 | ||||
| AK045345.1 | BAC32320.1 | |||
| AK045445.1 | BAC32372.1 | |||
| AK053314.1 | ||||
| AK082579.1 | BAC38536.1 | |||
| AK083878.1 | ||||
| AK159388.1 | BAE35043.1 | |||
| AK168935.1 | ||||
| BC011146.1 | AAH11146.1 |
More Information
Set Membership: |
|

Genomic Map