Assay Details
Target Gene Details
Entrez Gene ID: | 11634 |
Gene Name: | autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy) |
Gene Aliases: |
- |
Location: |
Chr.10:78030022-78043610 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Target Gene Details
Entrez Gene ID: | 54427 |
Gene Name: | DNA (cytosine-5-)-methyltransferase 3-like |
Gene Aliases: |
D6Ertd14e, ecat7 |
Location: |
Chr.10:78042287-78063622 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnmt3l | NM_001081695.2 | NP_001075164.1 | ||
| EF051624.1 |
More Information
Set Membership: |
|

Genomic Map